Gene Transcripts

Gene Symbol
UBE3C
Entrez Gene ID
9690
Gene Name
ubiquitin protein ligase E3C
Gene Aliases
HECTH2
Chromosome Location
Chr. 7: 157138952 - 157269372 on Build GRCh38
UniGene ID
Hs.118351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014671.2 NP_055486.2 11, 12 1735
XM_005249564.4 XP_005249621.1 10, 11 1669
XM_017012818.1 XP_016868307.1 12, 13 2007
GenBank mRNA AK295138.1 - 11, 12 1415
BC014029.2 - 12, 13 1495 4541634
D13635.1 - 11, 12 1726
DB085748.1 - 2, 3 265

View Full Product Details