Gene Transcripts

Gene Symbol
Best4
Entrez Gene ID
689103
Gene Name
bestrophin 4
Gene Aliases
Chromosome Location
Chr. 5: 136014017 - 136017742 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001066317.2 XP_001066317.1 6, 7 874

View Full Product Details