Gene Transcripts

Gene Symbol
Ankk1
Entrez Gene ID
367080
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
Chromosome Location
Chr. 8: 53750631 - 53758875 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766198.1 XP_008764420.1 8 1654

View Full Product Details