Gene Transcripts

Gene Symbol
ACAP1
Entrez Gene ID
9744
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 1
Gene Aliases
CENTB1
Chromosome Location
Chr. 17: 7336506 - 7354944 on Build GRCh38
UniGene ID
Hs.337242
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014716.3 NP_055531.1 22 2343
GenBank mRNA BC018543.1 - 22 2318
BT009788.1 - 22 2137
D30758.1 - 22 2325

View Full Product Details