Gene Transcripts

Gene Symbol
Lrr1
Entrez Gene ID
685860
Gene Name
leucine rich repeat protein 1
Gene Aliases
Ppil5
Chromosome Location
Chr. 6: 91463191 - 91472308 on Build Rnor_6.0
UniGene ID
Rn.198741
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109483.1 NP_001102953.1 4 1243
XM_008764719.1 XP_008762941.1 3 1261

View Full Product Details