Gene Transcripts

Gene Symbol
SPATA2
Entrez Gene ID
9825
Gene Name
spermatogenesis associated 2
Gene Aliases
PD1, PPP1R145, tamo
Chromosome Location
Chr. 20: 49903391 - 49915543 on Build GRCh38
UniGene ID
Hs.48513
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135773.1 NP_001129245.1 3 1869
NM_006038.3 NP_006029.1 3 1785
XM_006723894.1 XP_006723957.1 3 1705
XM_011529116.1 XP_011527418.1 3 1742
GenBank mRNA AB018300.1 - 3 1703
AK023229.1 - 3 1740
AK289644.1 - 3 1869
AK295536.1 - 2 1347
BC009481.2 - 3 1692 3953522
U28164.1 - 3 1750

View Full Product Details