Gene Transcripts

Gene Symbol
LOC691920
Entrez Gene ID
691920
Gene Name
similar to kinesin-like motor protein C20orf23
Gene Aliases
Chromosome Location
Chr. 3: 112428361 - 112506730 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001080099.5 XP_001080099.4 559
XM_006234815.2 XP_006234877.1 559

View Full Product Details