Gene Transcripts

Gene Symbol
Alyref
Entrez Gene ID
690585
Gene Name
Aly/REF export factor
Gene Aliases
Thoc4
Chromosome Location
Chr. 10: 109768599 - 109774639 on Build Rnor_6.0
UniGene ID
Rn.6994
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109602.1 NP_001103072.1 3 483
XM_006247925.2 XP_006247987.1 3 478

View Full Product Details