Gene Transcripts

Gene Symbol
Calr4
Entrez Gene ID
689537
Gene Name
calreticulin 4
Gene Aliases
Chromosome Location
Chr. 5: 128865250 - 128900135 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008763952.1 XP_008762174.1 7 1071

View Full Product Details