Gene Transcripts

Gene Symbol
ARHGAP25
Entrez Gene ID
9938
Gene Name
Rho GTPase activating protein 25
Gene Aliases
HEL-S-308, KAIA0053
Chromosome Location
Chr. 2: 68710505 - 68826833 on Build GRCh38
UniGene ID
Hs.531807
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007231.2 NP_001007232.2 10 1725
NM_001166276.1 NP_001159748.1 9 1665
NM_001166277.1 NP_001159749.1 9 1605
NM_014882.2 NP_055697.1 9 1662
XM_005264675.2 XP_005264732.1 10 1429
XM_005264676.2 XP_005264733.1 9 1313
XM_011533206.2 XP_011531508.1 10 1473
XM_011533207.2 XP_011531509.1 10 1439
XM_011533208.2 XP_011531510.1 11 1490
XM_011533209.2 XP_011531511.1 7 1030
XM_011533210.2 XP_011531512.1 5 557
XM_017005423.1 XP_016860912.1 10 1333
XM_017005424.1 XP_016860913.1 10 1330
XM_017005425.1 XP_016860914.1 9 1216
XM_017005426.1 XP_016860915.1 7 1667
XM_017005427.1 XP_016860916.1 7 1664
GenBank mRNA AK290396.1 - 10 1403
AK297056.1 - 9 1483
AK297157.1 - 10 1465
AK298669.1 - 9 1492
D29642.1 - 9 1475
EU794656.1 - 9 1662

View Full Product Details