Gene Transcripts

Gene Symbol
Btbd6
Entrez Gene ID
690367
Gene Name
BTB (POZ) domain containing 6
Gene Aliases
Chromosome Location
Chr. 6: 137785665 - 137788442 on Build Rnor_6.0
UniGene ID
Rn.6057
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077683.2 NP_001071151.2 4 1093
XM_006240664.2 XP_006240726.1 5 1247
GenBank mRNA BC127548.1 - 3 1021

View Full Product Details