Gene Transcripts

Gene Symbol
Stk33
Entrez Gene ID
690861
Gene Name
serine/threonine kinase 33
Gene Aliases
Chromosome Location
Chr. 1: 173872744 - 174051599 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759823.1 XP_008758045.1 4 827
XM_008759824.1 XP_008758046.1 3 532

View Full Product Details