Gene Transcripts

Gene Symbol
ARRDC1-AS1
Entrez Gene ID
85026
Gene Name
ARRDC1 antisense RNA 1
Gene Aliases
C9orf37
Chromosome Location
Chr. 9: 137615332 - 137618895 on Build GRCh38
UniGene ID
Hs.522520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_122035.1 3 1172
NR_122036.1 3 861
GenBank mRNA AF267857.1 - 3 1089
BC011781.2 - 3 1074
BC066646.1 - 3 879 5749061
BC117267.1 - 2 632 40125818

View Full Product Details