Gene Transcripts

Gene Symbol
C11orf94
Entrez Gene ID
143678
Gene Name
chromosome 11 open reading frame 94
Gene Aliases
Chromosome Location
Chr. 11: 45906534 - 45907282 on Build GRCh38
UniGene ID
Hs.193741
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080446.2 NP_001073915.2 2 187

View Full Product Details