Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005727.3 | NP_005718.2 | 7 | 1055 | |
XM_011540460.2 | XP_011538762.1 | 7 | 1029 | ||
GenBank mRNA | AF054838.1 | - | 5 | 581 | |
AF065388.1 | - | 6 | 702 | ||
AF133425.1 | - | 6 | 704 | ||
AK313774.1 | - | 7 | 1045 | ||
BC007290.1 | - | 6 | 708 | 3349295 | |
BC013404.2 | - | 6 | 706 | 2959379 |