Gene Transcripts

Gene Symbol
PHGR1
Entrez Gene ID
644844
Gene Name
proline, histidine and glycine rich 1
Gene Aliases
Chromosome Location
Chr. 15: 40351033 - 40356434 on Build GRCh38
UniGene ID
Hs.447537
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145643.1 NP_001139115.1 2 43
GenBank mRNA BC109367.1 - 1 14
BM854696.1 - 2 43

View Full Product Details