Gene Transcripts

Gene Symbol
SPACA5B
Entrez Gene ID
729201
Gene Name
sperm acrosome associated 5B
Gene Aliases
LYZL5B
Chromosome Location
Chr. X: 48130657 - 48132613 on Build GRCh38
UniGene ID
Hs.319319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079900.1 NP_001073369.1 1, 2 185

Gene Symbol
SPACA5
Entrez Gene ID
389852
Gene Name
sperm acrosome associated 5
Gene Aliases
LYC5, LYZL5, PNPK6288, SLLP-X, SLLP2, UNQ6288
Chromosome Location
Chr. X: 48004336 - 48009736 on Build GRCh38
UniGene ID
Hs.319319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_205856.3 NP_995328.2 2, 3 322
XM_017029512.1 XP_016885001.1 1, 2 937
GenBank mRNA AF217622.1 - 1, 2 178
AY358844.1 - 2, 3 322
BX279755.1 - 1, 2 171

View Full Product Details