Gene Transcripts
- Gene Symbol
- Mtpn
- Entrez Gene ID
- 14489
- Gene Name
- myotrophin
- Gene Aliases
- 5033418D15Rik, Gcdp, V1
- Chromosome Location
- Chr. 6: 35508824 - 35539888 on Build GRCm38
- UniGene ID
- Mm.182746
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008098.4 |
NP_032124.1
|
1, 2 |
359 |
|
GenBank mRNA |
AK028027.1 |
- |
1, 2 |
287 |
|
AK049398.1 |
- |
1, 2 |
259 |
|
AK083888.1 |
- |
1, 2 |
273 |
|
AK132029.1 |
- |
1, 2 |
282 |
|
AK145755.1 |
- |
1, 2 |
264 |
|
AK146305.1 |
- |
1, 2 |
307 |
|
AK146473.1 |
- |
1, 2 |
259 |
|
AK150750.1 |
- |
1, 2 |
281 |
|
AK151106.1 |
- |
1, 2 |
227 |
|
AK151147.1 |
- |
1, 2 |
227 |
|
AK151485.1 |
- |
1, 2 |
284 |
|
AK151564.1 |
- |
1, 2 |
227 |
|
AK151640.1 |
- |
1, 2 |
282 |
|
AK151687.1 |
- |
1, 2 |
255 |
|
AK152272.1 |
- |
1, 2 |
255 |
|
AK152317.1 |
- |
1, 2 |
245 |
|
BC043084.1 |
- |
1, 2 |
256 |
6417343 |
BC054811.1 |
- |
1, 2 |
272 |
6839739 |
CA559569.1 |
- |
1, 2 |
359 |
|
D78188.1 |
- |
1, 2 |
268 |
|
U20290.1 |
- |
1, 2 |
290 |
|
View Full Product Details