Gene Transcripts

Gene Symbol
TIMM17A
Entrez Gene ID
10440
Gene Name
translocase of inner mitochondrial membrane 17 homolog A (yeast)
Gene Aliases
TIM17, TIM17A
Chromosome Location
Chr. 1: 201955491 - 201970661 on Build GRCh38
UniGene ID
Hs.20716
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006335.2 NP_006326.1 3 193
GenBank mRNA AF106622.1 - 3 164
AK315602.1 - 3 181
AU123184.1 - 3 193
BC000294.2 - 3 163 2819455
BC004439.1 - 3 163 2819455
BC007106.1 - 3 174 4284092
BC009784.1 - 3 185 3683125
BC013823.1 - 3 152
BC015098.1 - 3 191 3686369
BC016817.1 - 1 48
BC020833.1 - 3 171 4773149
X97544.1 - 3 170

View Full Product Details