Gene Transcripts
- Gene Symbol
- Vmn2r41
- Entrez Gene ID
- 100042848
- Gene Name
- vomeronasal 2, receptor 41
- Gene Aliases
- Chromosome Location
- Chr. 7: 8137905 - 8161551 on Build GRCm38
- UniGene ID
- Mm.469892
- Species
- Mus musculus
- Gene Symbol
- Vmn2r35
- Entrez Gene ID
- 625353
- Gene Name
- vomeronasal 2, receptor 35
- Gene Aliases
- EG625353
- Chromosome Location
- Chr. 7: 7786151 - 7819867 on Build GRCm38
- UniGene ID
- Mm.469824
- Species
- Mus musculus
- Gene Symbol
- Vmn2r36
- Entrez Gene ID
- 100042653
- Gene Name
- vomeronasal 2, receptor 36
- Gene Aliases
- 100042724, Gm3991
- Chromosome Location
- Chr. 7: 7876653 - 7902462 on Build GRCm38
- UniGene ID
- Mm.465202
- Species
- Mus musculus
- Gene Symbol
- Vmn2r39
- Entrez Gene ID
- 545909
- Gene Name
- vomeronasal 2, receptor 39
- Gene Aliases
- EG545909
- Chromosome Location
- Chr. 7: 9014750 - 9030682 on Build GRCm38
- UniGene ID
- Mm.469894
- Species
- Mus musculus
- Gene Symbol
- Vmn2r40
- Entrez Gene ID
- 100042781
- Gene Name
- vomeronasal 2, receptor 40
- Gene Aliases
- Chromosome Location
- Chr. 7: 8907734 - 8931402 on Build GRCm38
- UniGene ID
- Mm.469893
- Species
- Mus musculus
- Gene Symbol
- Vmn2r43
- Entrez Gene ID
- 381838
- Gene Name
- vomeronasal 2, receptor 43
- Gene Aliases
- EC2-V2R, Gm1961
- Chromosome Location
- Chr. 7: 8244348 - 8260599 on Build GRCm38
- UniGene ID
- Mm.247383
- Species
- Mus musculus
- Gene Symbol
- Vmn2r48
- Entrez Gene ID
- 625580
- Gene Name
- vomeronasal 2, receptor 48
- Gene Aliases
- EG625580
- Chromosome Location
- Chr. 7: 9927624 - 9953585 on Build GRCm38
- UniGene ID
- Mm.469893
- Species
- Mus musculus
- Gene Symbol
- Vmn2r50
- Entrez Gene ID
- 434117
- Gene Name
- vomeronasal 2, receptor 50
- Gene Aliases
- EG434117
- Chromosome Location
- Chr. 7: 10037235 - 10053178 on Build GRCm38
- UniGene ID
- Mm.247383
- Species
- Mus musculus
View Full Product Details