Gene Transcripts

Gene Symbol
H2afj
Entrez Gene ID
232440
Gene Name
H2A histone family, member J
Gene Aliases
E130307C13
Chromosome Location
Chr. 6: 136808248 - 136810074 on Build GRCm38
UniGene ID
Mm.154564
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177688.4 NP_808356.1 1 1443
GenBank mRNA AK053755.1 - 1 1370

View Full Product Details