Gene Transcripts

Gene Symbol
Bola2
Entrez Gene ID
66162
Gene Name
bolA-like 2 (E. coli)
Gene Aliases
1110025L05Rik, BolA-2
Chromosome Location
Chr. 7: 126696000 - 126696693 on Build GRCm38
UniGene ID
Mm.358692
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175103.3 NP_780312.1 3 237
GenBank mRNA AK002662.1 - 3 240
AK003922.1 - 3 240
AK028165.1 - 3 251
AK140279.1 - 592
BC045523.1 - 3 255 6397886
BC061197.1 - 2 539 6707992

View Full Product Details