Gene Transcripts

Gene Symbol
USP19
Entrez Gene ID
10869
Gene Name
ubiquitin specific peptidase 19
Gene Aliases
ZMYND9
Chromosome Location
Chr. 3: 49108046 - 49120938 on Build GRCh38
UniGene ID
Hs.255596
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199160.1 NP_001186089.1 16 2618
NM_001199161.1 NP_001186090.1 16 2624
NM_001199162.1 NP_001186091.1 16 2588
NM_006677.2 NP_006668.1 15 2315
XM_005264823.2 XP_005264880.1 16 2625
XM_005264824.2 XP_005264881.1 16 2644
XM_005264825.2 XP_005264882.1 16 2619
XM_005264826.2 XP_005264883.1 16 2619
XM_005264827.2 XP_005264884.1 16 2580
XM_005264829.2 XP_005264886.1 16 2625
XM_005264830.2 XP_005264887.1 15 2322
XM_005264831.2 XP_005264888.1 15 2271
XM_006712946.2 XP_006713009.1 16 2622
XM_006712947.2 XP_006713010.1 16 2571
XM_006712948.2 XP_006713011.1 16 2613
XM_006712949.2 XP_006713012.1 16 2475
XM_006712950.2 XP_006713013.1 16 2574
XM_006712951.2 XP_006713014.1 16 2571
XM_006712952.2 XP_006713015.1 16 2568
XM_006712953.1 XP_006713016.1 16 2539
XM_017005615.1 XP_016861104.1 16 2616
XM_017005616.1 XP_016861105.1 16 2610
XM_017005617.1 XP_016861106.1 16 2574
XM_017005618.1 XP_016861107.1 16 2619
XM_017005619.1 XP_016861108.1 16 2616
XM_017005620.1 XP_016861109.1 16 2610
XM_017005621.1 XP_016861110.1 16 2469
XM_017005622.1 XP_016861111.1 16 2580
XM_017005623.1 XP_016861112.1 16 2463
XM_017005624.1 XP_016861113.1 16 2463
XM_017005625.1 XP_016861114.1 15 2316
XM_017005626.1 XP_016861115.1 15 2329
XM_017005627.1 XP_016861116.1 15 2277
XM_017005628.1 XP_016861117.1 15 2271
XM_017005629.1 XP_016861118.1 15 2386
XM_017005630.1 XP_016861119.1 15 2322
XM_017005631.1 XP_016861120.1 15 2316
XM_017005632.1 XP_016861121.1 15 2316
XM_017005633.1 XP_016861122.1 15 2310
XM_017005634.1 XP_016861123.1 15 2277
XM_017005635.1 XP_016861124.1 9 1053
GenBank mRNA AB020698.1 - 15 2157
AK294756.1 - 16 2511
AK300425.1 - 16 2442
BC014934.1 - 11 1596
BC065909.1 - 4 395
BC082241.1 - 13 1691
BC142660.1 - 16 2494
BC142727.1 - 16 2500
BC146752.1 - 15 2157 9007186

View Full Product Details