Gene Transcripts

Gene Symbol
YIF1A
Entrez Gene ID
10897
Gene Name
Yip1 interacting factor homolog A, membrane trafficking protein
Gene Aliases
54TM, FinGER7, YIF1, YIF1P
Chromosome Location
Chr. 11: 66284580 - 66289167 on Build GRCh38
UniGene ID
Hs.446445
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020470.2 NP_065203.2 6, 7 819
XM_005273720.3 XP_005273777.1 6, 7 795
XM_017017139.1 XP_016872628.1 6, 7 831
XM_017017140.1 XP_016872629.1 6, 7 831
GenBank mRNA AF004876.1 - 6, 7 752
AK226003.1 - 6, 7 797
AK291124.1 - 6, 7 797
BC001299.1 - 6, 7 770 3451489
BC009892.2 - 3, 4 246
CR456710.1 - 6, 7 637

View Full Product Details