Gene Transcripts

Gene Symbol
C1QB
Entrez Gene ID
713
Gene Name
complement C1q B chain
Gene Aliases
Chromosome Location
Chr. 1: 22653189 - 22661536 on Build GRCh38
UniGene ID
Hs.8986
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000491.3 NP_000482.3 3 471
XM_011542059.2 XP_011540361.1 4 622
GenBank mRNA BC008983.1 - 3 413
BQ711602.1 - 4 633
X03084.1 - 2 265

View Full Product Details