Gene Transcripts

Gene Symbol
SF3A3
Entrez Gene ID
10946
Gene Name
splicing factor 3a subunit 3
Gene Aliases
PRP9, PRPF9, SAP61, SF3a60
Chromosome Location
Chr. 1: 37956975 - 37990110 on Build GRCh38
UniGene ID
Hs.77897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320830.1 NP_001307759.1 12 1206
NM_006802.3 NP_006793.1 14 1365
GenBank mRNA AK128438.1 - 14 2174
AK222554.1 - 14 1241
AK291181.1 - 14 1246
AK292116.1 - 14 1276
AK301424.1 - 12 1083
BC002395.2 - 14 1258 2821350
BC011523.1 - 14 1322 3867442
BM455074.1 - 4 397
BM905122.1 - 1 53
N91991.1 - 2 95
U08815.1 - 14 1228

View Full Product Details