Gene Transcripts

Gene Symbol
STIP1
Entrez Gene ID
10963
Gene Name
stress induced phosphoprotein 1
Gene Aliases
HEL-S-94n, HOP, IEF-SSP-3521, P60, STI1, STI1L
Chromosome Location
Chr. 11: 64185272 - 64204548 on Build GRCh38
UniGene ID
Hs.337295
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282652.1 NP_001269581.1 13 2215
NM_001282653.1 NP_001269582.1 13 1596
NM_006819.2 NP_006810.1 13 1668
GenBank mRNA AK291555.1 - 13 1595
AK297319.1 - 13 1507
BC002987.1 - 13 1583 2823157
BC039299.1 - 13 2215 4830381
BT020010.1 - 12 1521
BT020011.1 - 12 1521
CR536512.1 - 12 1521
FJ224350.1 - 13 1583
M86752.1 - 13 1583

View Full Product Details