Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001126335.1 | NP_001119807.1 | 11 | 1300 | |
NM_001243036.1 | NP_001229965.1 | 11 | 1395 | ||
NM_014270.4 | NP_055085.1 | 11 | 1386 | ||
XM_006722992.1 | XP_006723055.1 | 9 | 1061 | ||
XM_011526402.2 | XP_011524704.1 | 11 | 2490 | ||
XM_017026230.1 | XP_016881719.1 | 9 | 1036 | ||
GenBank mRNA | AB033548.1 | - | 11 | 1271 | |
AF141289.1 | - | 11 | 1368 | ||
AJ249199.1 | - | 10 | 1245 | ||
AK026446.1 | - | 11 | 1369 | ||
AK223147.1 | - | 11 | 1374 | ||
AK313708.1 | - | 11 | 1287 | ||
AL365340.1 | - | 4 | 438 | ||
AL365341.1 | - | 3 | 222 | ||
AY170373.1 | - | 6 | 717 | ||
BC017962.1 | - | 11 | 1375 | 4608486 |