Gene Transcripts

Gene Symbol
Vps37c
Entrez Gene ID
308178
Gene Name
vacuolar protein sorting 37 homolog C (S. cerevisiae)
Gene Aliases
RGD1309258
Chromosome Location
Chr. 1: 226742274 - 226767868 on Build Rnor_6.0
UniGene ID
Rn.12915
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107463.1 NP_001100933.1 5 638
XM_006231033.2 XP_006231095.1 5 696
XM_006231034.2 XP_006231096.1 5 700
XM_006231035.2 XP_006231097.1 5 1662
GenBank mRNA BC169040.1 - 5 631

Gene Symbol
LOC100910929
Entrez Gene ID
100910929
Gene Name
vacuolar protein sorting-associated protein 37C-like
Gene Aliases
Chromosome Location
Chr. Un|NW_007906915: 116348 - 141939 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749059.3 XP_003749107.1 5 696
XM_006231078.2 XP_006231140.1 5 700
XM_006231079.2 XP_006231141.1 5 1662

View Full Product Details