Gene Transcripts

Gene Symbol
Tiprl
Entrez Gene ID
360869
Gene Name
TOR signaling pathway regulator
Gene Aliases
RGD1310442
Chromosome Location
Chr. 13: 83478966 - 83504448 on Build Rnor_6.0
UniGene ID
Rn.56867
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109667.1 NP_001103137.1 3, 4 431
GenBank mRNA BC128780.1 - 3, 4 431

View Full Product Details