Gene Transcripts

Gene Symbol
Cib1
Entrez Gene ID
81823
Gene Name
calcium and integrin binding 1 (calmyrin)
Gene Aliases
Cib, Sip2-28
Chromosome Location
Chr. 1: 142014962 - 142020461 on Build Rnor_6.0
UniGene ID
Rn.833
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031145.1 NP_112407.1 6 524
GenBank mRNA AF136585.1 - 6 524
BC091143.1 - 6 510 7308828

View Full Product Details