Gene Transcripts

Gene Symbol
AAK1
Entrez Gene ID
22848
Gene Name
AP2 associated kinase 1
Gene Aliases
Chromosome Location
Chr. 2: 69457995 - 69643845 on Build GRCh38
UniGene ID
Hs.468878
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014911.3 NP_055726.3 3 705
GenBank mRNA AB028971.1 - 3 584
AK056043.1 - 3 512
AK310951.1 - 3 508
BC002695.2 - 2 240
BC104842.1 - 2 224 8143845
BC111965.1 - 2 224 8327433
BC143710.1 - 2 224 9052224
DB285703.1 - 3 513

View Full Product Details