Gene Transcripts
- Gene Symbol
- CA4
- Entrez Gene ID
- 762
- Gene Name
- carbonic anhydrase 4
- Gene Aliases
- CAIV, Car4, RP17
- Chromosome Location
- Chr. 17: 60149941 - 60178118 on Build GRCh38
- UniGene ID
- Hs.89485
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_005257639.2 |
XP_005257696.1
|
4, 5 |
515 |
|
XM_011525183.2 |
XP_011523485.1
|
4, 5 |
504 |
|
XM_017025012.1 |
XP_016880501.1
|
4, 5 |
504 |
|
GenBank mRNA |
AK289715.1 |
- |
4, 5 |
514 |
|
BC057792.1 |
- |
4, 5 |
520 |
30331755 |
BC069649.1 |
- |
4, 5 |
467 |
7262441 |
BC074768.2 |
- |
4, 5 |
444 |
30915189 |
CR541766.1 |
- |
4, 5 |
421 |
|
DA113846.1 |
- |
4, 5 |
515 |
|
M83670.1 |
- |
4, 5 |
468 |
|
View Full Product Details