Gene Transcripts
- Gene Symbol
- C17orf49
- Entrez Gene ID
- 124944
- Gene Name
- chromosome 17 open reading frame 49
- Gene Aliases
- BAP18, HEPIS
- Chromosome Location
- Chr. 17: 7014737 - 7017524 on Build GRCh38
- UniGene ID
- Hs.511801
- Species
- Homo sapiens
- Gene Symbol
- RNASEK-C17orf49
- Entrez Gene ID
- 100529209
- Gene Name
- RNASEK-C17orf49 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 17: 7012417 - 7017524 on Build GRCh38
- UniGene ID
- Hs.511801
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037717.1 |
|
5 |
1264 |
|
GenBank mRNA |
BC040148.1 |
- |
5 |
1021 |
|
View Full Product Details