Gene Transcripts

Gene Symbol
MTPN
Entrez Gene ID
136319
Gene Name
myotrophin
Gene Aliases
GCDP, V-1
Chromosome Location
Chr. 7: 135926755 - 135977456 on Build GRCh38
UniGene ID
Hs.602015
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145808.3 NP_665807.1 1 403
GenBank mRNA AF106075.1 - 1 252
AK056225.1 - 1 306
AK309816.1 - 1 301
AL834231.1 - 1 275
BC028093.1 - 1 301 3876632
DA325130.1 - 1 403

Gene Symbol
LUZP6
Entrez Gene ID
767558
Gene Name
leucine zipper protein 6
Gene Aliases
MPD6, MTPNUT
Chromosome Location
Chr. 7: 135926755 - 135977456 on Build GRCh38
UniGene ID
Hs.602015
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128619.2 NP_001122091.2 1 403
GenBank mRNA DA325904.1 - 1 325

View Full Product Details