Gene Transcripts

Gene Symbol
C6orf1
Entrez Gene ID
221491
Gene Name
chromosome 6 open reading frame 1
Gene Aliases
LBH
Chromosome Location
Chr. 6: 34246380 - 34249108 on Build GRCh38
UniGene ID
Hs.381300
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008703.2 NP_001008703.1 5 502
NM_001008704.2 NP_001008704.1 5 442
NM_001287396.1 NP_001274325.1 5 694
NM_001287397.1 NP_001274326.1 4 728
NM_001287398.1 NP_001274327.1 4 476
NM_178508.4 NP_848603.2 5 754
GenBank mRNA AK290164.1 - 5 436
BC023627.2 - 5 374 4764944
BC047919.1 - 5 635 6190218
BM474526.1 - 4 393
BU194178.1 - 5 575
BU956518.1 - 4 241
CK001844.1 - 5 505

View Full Product Details