Gene Transcripts

Gene Symbol
DSCR9
Entrez Gene ID
257203
Gene Name
Down syndrome critical region 9 (non-protein coding)
Gene Aliases
NCRNA00038
Chromosome Location
Chr. 21: 37208503 - 37221736 on Build GRCh38
UniGene ID
Hs.505159
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026719.2 3 1617
GenBank mRNA AB066100.1 - 1 1384
BC029827.1 - 1 955
BC066653.1 - 3 1617

View Full Product Details