Gene Transcripts
- Gene Symbol
- APOC4
- Entrez Gene ID
- 346
- Gene Name
- apolipoprotein C4
- Gene Aliases
- APO-CIV, APOC-IV
- Chromosome Location
- Chr. 19: 44942238 - 44945496 on Build GRCh38
- UniGene ID
- Hs.731599
- Species
- Homo sapiens
- Gene Symbol
- APOC4-APOC2
- Entrez Gene ID
- 100533990
- Gene Name
- APOC4-APOC2 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 19: 44942238 - 44949565 on Build GRCh38
- UniGene ID
- Hs.731599
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037932.1 |
|
2 |
244 |
|
GenBank mRNA |
AK126257.1 |
- |
2 |
244 |
|
View Full Product Details