Gene Transcripts

Gene Symbol
PDE4DIP
Entrez Gene ID
9659
Gene Name
phosphodiesterase 4D interacting protein
Gene Aliases
CMYA2, MMGL
Chromosome Location
Chr. 1: 148808167 - 149051277 on Build GRCh38
UniGene ID
Hs.584841
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198832.2 NP_001185761.2 30 4311
NM_001198834.3 NP_001185763.3 27 4359
NM_014644.5 NP_055459.5 27 4359
XM_005272981.3 XP_005273038.2 23 4986
XM_005277442.4 XP_005277499.2 23 4986
XM_005277443.3 XP_005277500.1 23 4986
XM_006711642.2 XP_006711705.1 23 4986
XM_006711643.2 XP_006711706.1 23 4983
XM_006711644.2 XP_006711707.1 23 4986
XM_006711646.1 XP_006711709.1 30 4648
XM_006711648.2 XP_006711711.1 23 4656
XM_006711650.3 XP_006711713.1 28 4133
XM_006711651.1 XP_006711714.1 27 4251
XM_006711652.1 XP_006711715.1 27 4248
XM_011510172.1 XP_011508474.1 30 4806
XM_011510173.1 XP_011508475.1 30 4803
XM_011510174.1 XP_011508476.1 23 4986
XM_011510175.1 XP_011508477.1 30 4803
XM_011510176.1 XP_011508478.1 30 4476
XM_011510179.1 XP_011508481.1 27 4248
XM_017002875.1 XP_016858364.1 23 4986
XM_017002876.1 XP_016858365.1 23 4986
XM_017002877.1 XP_016858366.1 23 4983
XM_017002878.1 XP_016858367.1 30 4806
XM_017002879.1 XP_016858368.1 23 4986
XM_017002880.1 XP_016858369.1 30 4806
XM_017002881.1 XP_016858370.1 23 4986
XM_017002882.1 XP_016858371.1 23 4983
XM_017002883.1 XP_016858372.1 23 4986
XM_017002884.1 XP_016858373.1 23 4986
XM_017002885.1 XP_016858374.1 23 4656
XM_017002886.1 XP_016858375.1 23 4653
XM_017002887.1 XP_016858376.1 31 5083
XM_017002888.1 XP_016858377.1 31 5056
XM_017002889.1 XP_016858378.1 31 4728
XM_017002890.1 XP_016858379.1 23 4986
XM_017002891.1 XP_016858380.1 29 4887
XM_017002892.1 XP_016858381.1 29 4518
XM_017002893.1 XP_016858382.1 30 5006
XM_017002894.1 XP_016858383.1 30 4651
XM_017002895.1 XP_016858384.1 27 3969
XM_017002896.1 XP_016858385.1 23 4986
XM_017002897.1 XP_016858386.1 23 4983
XM_017002898.1 XP_016858387.1 30 4648
XM_017002899.1 XP_016858388.1 30 4318
XM_017002900.1 XP_016858389.1 23 4656
XM_017002901.1 XP_016858390.1 23 4656
XM_017002902.1 XP_016858391.1 30 4315
XM_017002903.1 XP_016858392.1 30 4318
GenBank mRNA AB007923.2 - 31 4190
BX647660.1 - 27 4287
DA238288.1 - 4 552

Gene Symbol
LOC100996724
Entrez Gene ID
100996724
Gene Name
phosphodiesterase 4D interacting protein-like
Gene Aliases
Chromosome Location
Chr. 1: 120471449 - 120701470 on Build GRCh38
UniGene ID
Hs.731111
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017003064.1 XP_016858553.1 28 4223
GenBank mRNA CR749273.1 - 8 1491

View Full Product Details