Gene Transcripts

Gene Symbol
ANKRD53
Entrez Gene ID
79998
Gene Name
ankyrin repeat domain 53
Gene Aliases
Chromosome Location
Chr. 2: 70977183 - 70985499 on Build GRCh38
UniGene ID
Hs.512744
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024933.3 NP_079209.3 6 1123
GenBank mRNA BC035234.1 - 6 1123

View Full Product Details