Gene Transcripts
- Gene Symbol
- KIAA1549L
- Entrez Gene ID
- 25758
- Gene Name
- KIAA1549 like
- Gene Aliases
- C11orf41, C11orf69, G2
- Chromosome Location
- Chr. 11: 33376569 - 33674102 on Build GRCh38
- UniGene ID
- Hs.502266
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012194.2 |
NP_036326.2
|
5, 6 |
3081 |
|
XM_005252847.3 |
XP_005252904.2
|
7, 8 |
3175 |
|
XM_005252848.3 |
XP_005252905.1
|
5, 6 |
3099 |
|
XM_011519969.2 |
XP_011518271.1
|
5, 6 |
3099 |
|
XM_011519970.2 |
XP_011518272.1
|
6, 7 |
1741 |
|
XM_017017484.1 |
XP_016872973.1
|
5, 6 |
3099 |
|
XM_017017485.1 |
XP_016872974.1
|
5, 6 |
3099 |
|
XM_017017486.1 |
XP_016872975.1
|
6, 7 |
1741 |
|
GenBank mRNA |
AK124395.1 |
- |
5, 6 |
3099 |
|
U10991.1 |
- |
5, 6 |
2488 |
|
View Full Product Details