Gene Transcripts

Gene Symbol
Stap1
Entrez Gene ID
56792
Gene Name
signal transducing adaptor family member 1
Gene Aliases
AI586015, Brdg1, STAP-1
Chromosome Location
Chr. 5: 86071585 - 86104784 on Build GRCm38
UniGene ID
Mm.131237
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019992.3 NP_064376.1 1, 2 135
GenBank mRNA AB036058.1 - 1, 2 125
AK041474.1 - 1, 2 136
AK042475.1 - 1, 2 182
AK153661.1 - 1, 2 214
AK162655.1 - 1, 2 217
BC057898.1 - 1, 2 189
BC145550.1 - 1, 2 151 9054114
BC145828.1 - 1, 2 150 40131131
BC145830.1 - 1, 2 151 40131133

View Full Product Details