Gene Transcripts

Gene Symbol
Vmn2r6
Entrez Gene ID
667069
Gene Name
vomeronasal 2, receptor 6
Gene Aliases
EG620718, EG667069
Chromosome Location
Chr. 3: 64537501 - 64565434 on Build GRCm38
UniGene ID
Mm.484956
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104619.1 NP_001098089.1 1 256

View Full Product Details