Gene Transcripts
- Gene Symbol
- Vmn2r84
- Entrez Gene ID
- 625068
- Gene Name
- vomeronasal 2, receptor 84
- Gene Aliases
- EG625068
- Chromosome Location
- Chr. 10: 130385800 - 130394241 on Build GRCm38
- UniGene ID
- Mm.359168
- Species
- Mus musculus
- Gene Symbol
- A330087I24
- Entrez Gene ID
- 327857
- Gene Name
- uncharacterized protein A330087I24
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.453020
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK039689.1 |
- |
5 |
1281 |
|
- Gene Symbol
- Vmn2r-ps104
- Entrez Gene ID
- 100041915
- Gene Name
- vomeronasal 2, receptor, pseudogene 104
- Gene Aliases
- Chromosome Location
- Chr. 13: 66351314 - 66355797 on Build GRCm38
- UniGene ID
- Mm.390608
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK049221.1 |
- |
|
1113 |
|
- Gene Symbol
- Vmn2r86
- Entrez Gene ID
- 625109
- Gene Name
- vomeronasal 2, receptor 86
- Gene Aliases
- EG625109
- Chromosome Location
- Chr. 10: 130446199 - 130455894 on Build GRCm38
- UniGene ID
- Mm.469845
- Species
- Mus musculus
View Full Product Details