Gene Transcripts

Gene Symbol
Tnfsfm13
Entrez Gene ID
619441
Gene Name
tumor necrosis factor (ligand) superfamily, membrane-bound member 13
Gene Aliases
Tnfsf12-Tnfsf13, Tnfsf12Tnfsf13, Tnfsf12tnfsf13, Twe-pril
Chromosome Location
Chr. 11: 69682577 - 69696098 on Build GRCm38
UniGene ID
Mm.8983
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034097.2 NP_001029269.1 13 1523
NM_001159503.2 NP_001152975.1 13 1517
GenBank mRNA AK044387.1 - 13 1524
BC096441.1 - 13 1229 3482756

Gene Symbol
Tnfsf13
Entrez Gene ID
69583
Gene Name
tumor necrosis factor (ligand) superfamily, member 13
Gene Aliases
2310026N09Rik, April, Tall2, Trdl1
Chromosome Location
Chr. 11: 69682577 - 69685554 on Build GRCm38
UniGene ID
Mm.8983
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159505.1 NP_001152977.1 6 964
NM_023517.2 NP_076006.2 6 967
GenBank mRNA AK009514.1 - 6 969
AK040667.1 - 6 742
AK134028.1 - 6 670
AK170351.1 - 6 971
BC069900.1 - 6 926 5290965

View Full Product Details