Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098782.1 | NP_001092252.1 | 2 | 91 | |
XM_006234861.2 | XP_006234923.1 | 2 | 162 | ||
GenBank mRNA | BC081918.1 | - | 2 | 91 | |
FQ213650.1 | - | 2 | 112 | ||
FQ216871.1 | - | 2 | 112 | ||
FQ217248.1 | - | 2 | 93 | ||
FQ217695.1 | - | 2 | 84 | ||
FQ222385.1 | - | 2 | 76 |