Gene Transcripts

Gene Symbol
Vom2r16
Entrez Gene ID
690691
Gene Name
vomeronasal 2 receptor, 16
Gene Aliases
Chromosome Location
Chr. 1: 62837021 - 62855155 on Build Rnor_6.0
UniGene ID
Rn.198000
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099655.1 NP_001093125.1 2 312

View Full Product Details