Gene Transcripts
- Gene Symbol
- ACSL6
- Entrez Gene ID
- 23305
- Gene Name
- acyl-CoA synthetase long-chain family member 6
- Gene Aliases
- ACS2, FACL6, LACS 6, LACS2, LACS5
- Chromosome Location
- Chr. 5: 131949973 - 132012068 on Build GRCh38
- UniGene ID
- Hs.14945
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001009185.2 |
NP_001009185.1
|
20, 21 |
2134 |
|
NM_001205247.1 |
NP_001192176.1
|
20, 21 |
2029 |
|
NM_001205248.1 |
NP_001192177.1
|
20, 21 |
2121 |
|
NM_001205250.1 |
NP_001192179.1
|
20, 21 |
2131 |
|
NM_001205251.1 |
NP_001192180.1
|
19, 20 |
1896 |
|
NM_015256.3 |
NP_056071.2
|
20, 21 |
2134 |
|
GenBank mRNA |
AB020644.1 |
- |
20, 21 |
2094 |
|
AF129166.1 |
- |
20, 21 |
2064 |
|
AK122702.1 |
- |
9, 10 |
1592 |
|
AK294291.1 |
- |
20, 21 |
2004 |
|
AK314452.1 |
- |
20, 21 |
2036 |
|
BC026161.1 |
- |
20, 21 |
2030 |
4830357 |
BC047453.1 |
- |
19, 20 |
1818 |
5270879 |
DQ083031.1 |
- |
20, 21 |
1995 |
|
View Full Product Details