Gene Transcripts

Gene Symbol
Samd15
Entrez Gene ID
681908
Gene Name
sterile alpha motif domain containing 15
Gene Aliases
Chromosome Location
Chr. 6: 111223026 - 111241326 on Build Rnor_6.0
UniGene ID
Rn.217378
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127609.1 NP_001121081.1 1 1429
XM_006240398.2 XP_006240460.1 1 1237
GenBank mRNA BC161884.1 - 1 1429

View Full Product Details