Gene Transcripts

Gene Symbol
Ap3b2
Entrez Gene ID
308777
Gene Name
adaptor-related protein complex 3, beta 2 subunit
Gene Aliases
Chromosome Location
Chr. 1: 143284041 - 143315846 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748891.3 XP_003748939.1 23 3130
XM_006229468.2 XP_006229530.1 23 3164
XM_006229469.2 XP_006229531.1 23 3135

View Full Product Details